Optical plate
Lieferant: DIDALAB
POD010240
DIDAPOD010240EA 0 CHF
DIDAPOD010240
Optical plate
Lehrmittel Physik-Lehrmittel Lehrmittel für die Optik
Optical glass components are manufactured with high accuracy polishing and flatness tolerance.
  • Optical polishing
  • Two sides with parallelism less than 1 µm