CCD camera, POD 010 020
Lieferant: DIDALAB
POD010020
DIDAPOD010020EA 1150 CHF
DIDAPOD010020
CCD camera, POD 010 020
Kameras
The CCD Caliens camera is designed with an optical head directly interfaced with a PC computer through USB port. It can sample and operate luminous pictures with a complete and easy-to-use software.
  • Sensor composed of 2048 pixels, 14 µm width (for about 30 mm)
  • Optical head, fitted out with stainless steel rod, Ø 10 mm, can be adapted to optical bench
Suitable for experiments like Fresnel's biprism, Gaussian beams, sodium doublets, source coherence and filter spectral width.
Spectrometry by Fourier's transforms with Michelson interferometer: WhenMore Product Information
Order Now
TECHNISCHE DATEN
Environmentally Preferable
Auflösung 2048 pixels
Belichtungszeit 5 ms to 0,1 s