4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride, fluorogenic, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyms: 5,6-Diamino-1,3-benzodioxole dihydrochloride, 1,3-Benzodioxole-5,6-diamine dihydrochloride, 1,2-Diamino-4,5-methylenedioxybenzene dihydrochloride, MDB, Benzo[1,3]dioxole-5-6-diamine dihydrochloride

MQ100

D4784-100MG D4784-10MG D4784-50MG
D4784-100MGEA 1490 CHF
D4784-100MG D4784-10MG D4784-50MG
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride, fluorogenic, Sigma-Aldrich®
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride is a fluorescence tag for α keto acids. It is also required for the derivatisation of sialic acids in presence of dilute sulfuric acid. The method allows determination of sialic acids.


4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride has been used for the derivatisation of sialic acids to analyse them by capillary liquid-chromatography-electrospray ionisation/tandem mass spectrometry (CapLC-ESI-MS/MS). Fluorogenic reagent for α-ketoacids.

Formel: C₇H₈N₂O₂·2HCl
MW: 225.07 g/mol
Melting Pt: 247 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00037497
CAS-Nummer: 81864-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da814b25819deeccdecacde0d061e15a4c3cc6de&vid=da814b25819deeccdecacde0d061e15a4c3cc6de&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Colour) White to Orange to Red
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon 36.4 - 38.3%
Nitrogen 12.0 - 12.8%
Purity (HPLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR