N,N-Dimethyl-4-nitrosoanilin 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyms: 4-Nitroso-N,N-dimethylaniline

M-Clarity™ quality level = MQ100

D172405-100G D172405-25G
D172405-100GEA 184 CHF
D172405-100G D172405-25G
N,N-Dimethyl-4-nitrosoanilin 97%, Sigma-Aldrich®
N,N-Dimethyl-4-nitrosoanilin

N,N-Dimethyl-4-nitrosoaniline has been used along with imidazole to detect the formation of singlet oxygen in cells.


N,N-Dimethyl-4-nitrosoaniline is used for the detection of singlet oxygen. Singlet oxygen reacts with imidazole and thereby bleaches N,N-Dimethyl-4-nitrosoaniline via oxidation. Bleaching of N,N-Dimethyl-4-nitrosoaniline is observed as reduction in the absorption at 438 nm.

Formel: C₈H₁₀N₂O
Siedepunkt: 259 °C (1013 hPa)
Melting Pt: 85...87 °C (lit.)
Dichte: 1.145 g/cm³ (20 °C)
Lager Temperatur: Room Temperature
MDL Number: MFCD00002063
CAS-Nummer: 138-89-6
EINECS: 205-343-1
UN: 1369
ADR: 4.2,II
Merck Index: 12,06736
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07acb402f41d42fce0b8e4d555d2ff8ed2f064f5&vid=2f144eb30aa4e5fd1878348e58d31685eaf10be5&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Form) Powder and/or Chunks
Conforms to Requirements
Infrared spectrum Conforms to Structure
Carbon 61.7 - 66.2 %
Nitrogen 18.0 - 19.3 %
Purity (TLC) ≥97 %
Solubility (Turbidity) Clear to Turbid
Solubility (Colour)
c = 5%, Ethanol
Green to Very Dark Green


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR