Trigger2Trigger lanyard, DBI-SALA®, 3M™
Lieferant: CAPITAL SAFETY
1500054
CPSD1500054EA 0 CHF
CPSD1500054
Trigger2Trigger lanyard, DBI-SALA®, 3M™
Fallschutz Verbindungsmittel und Seile
DBI-SALA® Trigger2Trigger lanyard is an economical tethering solution for most hand tools which is most commonly used in combination with DBI-SALA® attachment points. Pull down on the trigger to open the dual-hooked connector, then release once latched to a tool.
  • Available in several lengths
  • Trigger snaps swivel provides tangle-resistant
  • Thumb-controlled trigger snaps on both sides of these lanyard makes easy to operate, even while wearing gloves.
Capacity: 4,5 kg