GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
Lieferant: CUSTOM MADE CHEMICALS LAB
109500.L1
CMPL109500.L1EA 0 CHF
CMPL109500.L1
GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
Standards Kationischer Standards Mehrelement-Standards
16 components: Ag 10 mg/l, Al 100 mg/l, As 100 mg/l, Ba 50 mg/l, Be 5 mg/l, Cd 5 mg/l, Co 50 mg/l, Cr 20 mg/l, Cu 50 mg/l, Fe 20 mg/l, Mn 20 mg/l, Ni 50 mg/l, Pb 100 mg/l, Sb 100 mg/l, Se 100 mg/l, Tl 100 mg/l in 5% HNO₃.
Shelf life upto 36 Months, Store under normal laboratory conditions, at temperatures between 15 °C to 25 °C.
Zertifizierungen: ISO 17025/17034/9001