Ampicillin Trihydrat

Lieferant: Chemicon
Achtung

Synonyms: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

S4601
CHMCS4601EA 0 CHF
CHMCS4601
Ampicillin Trihydrat
Ampicillin Trihydrat
Formel: C₁₆H₁₉N₃O₄S·3H₂O
MW: 403.45 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD00072036
CAS-Nummer: 7177-48-2
EINECS: 200-709-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR