Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin)

Lieferant: Chemicon

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine

AB5822
CHMCAB5822EA 0 CHF
CHMCAB5822
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin)
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin)
Formel: C₁₀H₂₆N₄
MW: 202.35 g/mol
MDL Number: MFCD00008215
CAS-Nummer: 71-44-3
EINECS: 200-754-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR