Euro pallets and sumps, PE
Lieferant: CEMO
8276 8277 8278 8279 8280 8281 8282 8283 8284 8285 8286 8287 8432
CEMG8276EA 326 CHF
CEMG8276 CEMG8277 CEMG8278 CEMG8279 CEMG8280 CEMG8281 CEMG8282 CEMG8283 CEMG8284 CEMG8285 CEMG8286 CEMG8287 CEMG8432
Euro pallets and sumps, PE
Fasslager
These sump pallets offer protection and a clean environment, through high resistance against acids and lyes and are used for the storage of caustic, flammable, explosive, firepromoting or toxic substances. Manufactured from high quality polyethylene (HDPE). For storing drums up to 224 L.
  • High functionality and high chemical resistance
  • Tray can be placed directly on the floor or on a Euro pallet
  • Available in different versions with locking pins (for Euro-pallets), feet, runners or castors
Euro PE sump pallet (250/2 - RG) without approval no. areMore Product Information