Indometacin

Lieferant: Cayman Chemical

70270-1
CAYM70270-1EA 53.4 CHF
CAYM70270-1
Indometacin
Indomethacin
Formel: C₁₉H₁₆ClNO₄
MW: 357.80 g/mol
MDL Number: MFCD00057095
CAS-Nummer: 53-86-1
EINECS: 200-186-5
UN: 2811
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR