4-(2-Hydroxyethyl)phenol ≥98%

Lieferant: Cayman Chemical
Achtung

Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol

27600-100 27600-250 27600-500
CAYM27600-100EA 189 CHF
CAYM27600-100 CAYM27600-250 CAYM27600-500
4-(2-Hydroxyethyl)phenol ≥98%
2-(4-Hydroxyphenyl)ethanol

Tyrosol is a phenol that has been found in olives and olive oil and has diverse biological activities.


Bitte beachten This product is not for human or veterinary use.

Formel: C₈H₁₀O₂
MW: 138.17 g/mol
Siedepunkt: 310
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00002902
CAS-Nummer: 501-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR