Cholesterol margarate 95

Lieferant: Cayman Chemical

Synonyms: Cholesterol Heptadecanoate, Heptadecanoic Acid Cholesterol Ester, Margaric Acid Cholesterol Ester

21750-1 21750-100 21750-50 21750-500
CAYM21750-1EA 556 CHF
CAYM21750-1 CAYM21750-100 CAYM21750-50 CAYM21750-500
Cholesterol margarate 95
Cholesterol margarate

Cholesteryl heptadecanoate is a cholesterol ester (CE) formed by the condensation of cholesterol with heptadecanoic acid, a C-17 saturated fatty acid that does not occur in any natural animal or vegetable fat at high concentrations.


Used as an internal standard for the quantification of cholesterol esters by GC- or LC-mass spectrometry. CEs are major constituents of lipoprotein particles carried in blood and accumulate in the fatty acid lesions of atherosclerotic plaques. CEs of various fatty acids are major constituents of murine and human adrenal glands.

Formel: C₄₄H₇₈O₂
MW: 639.11 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00056438
CAS-Nummer: 24365-37-5

Order Now

Spezifikation Testergebnisse

Purity ≥95%
λmax 201 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR