BAM15 98, crystalline solid

Lieferant: Cayman Chemical

Synonyms: BAM 15, N5,N6-Bis(2-fluorophenyl)-[1,2,5]oxadiazolo[3,4-b]pyrazine-5,6-diamine

17811-1 17811-10 17811-25 17811-5
CAYM17811-1EA 65.5 CHF
CAYM17811-1 CAYM17811-10 CAYM17811-25 CAYM17811-5
BAM15 98, crystalline solid
BAM15

BAM15 is a mitochondrial protonophore uncoupler which increase proton transport into the mitochondrial matrix through an ATP synthase-independent pathway, thereby uncoupling nutrient oxidation from ATP production.


  • High oxygen flux than FCCP


These are used to determine rate of cellular respiration and have antioxidant effects that protect from ischemia-reperfusion injury. The off target activity of this uncouplers leads to the undesired effects, including plasma membrane depolarization, mitochondrial inhibition, and cytotoxicity.

Formel: C₁₆H₁₀F₂N₆O
MW: 340.29 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00373912
CAS-Nummer: 210302-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR