Anti-Nod2 Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
160777-1
CAYM160777-1EA 637 CHF
CAYM160777-1
Anti-Nod2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nod2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR