Anti-iPLA2 Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
160507-1
CAYM160507-1EA 637 CHF
CAYM160507-1
Anti-iPLA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: iPLA2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Hamster, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR