3-Brom-5-phenylsalicylsäure

Lieferant: Cayman Chemical

Synonyms: NSC 109116, 5-PBSA, 5-Bromo-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid, 5-Bromo-4-hydroxy[1,1'-biphenyl]-3-carboxylic acid, AKR1C1-IN-1

13574-1 13574-5 13574-10 13574-25
CAYM13574-1EA 133 CHF
CAYM13574-1 CAYM13574-5 CAYM13574-10 CAYM13574-25
3-Brom-5-phenylsalicylsäure
3-Brom-5-phenylsalicylsäure
Formel: C₁₃H₉BrO₃
MDL Number: MFCD18378258
CAS-Nummer: 4906-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR