Natriumbutyrat

Lieferant: Cayman Chemical

Synonyms: Butyric acid sodium salt, Sodium Butyrate

13121-50 13121-100 13121-250
CAYM13121-50EA 65.5 CHF
CAYM13121-50 CAYM13121-100 CAYM13121-250
Natriumbutyrat
Natriumbutyrat
Formel: C₄H₇NaO₂
Melting Pt: 244 °C
Dichte: 1.324 g/cm³ (30 °C)
MDL Number: MFCD00002816
CAS-Nummer: 156-54-7
EINECS: 205-857-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR