2-Amino-5-methyl-4-phenylthiazol

Lieferant: Cayman Chemical

13061-100 13061-250 13061-500 13061-1
CAYM13061-100EA 91.4 CHF
CAYM13061-100 CAYM13061-250 CAYM13061-500 CAYM13061-1
2-Amino-5-methyl-4-phenylthiazol
2-Amino-5-methyl-4-phenylthiazole
Formel: C₁₀H₁₀N₂S
MW: 190.2648 g/mol
MDL Number: MFCD00617257
CAS-Nummer: 30709-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR