Aloe emodin 95, crystalline solid

Lieferant: Cayman Chemical
Achtung

Synonyms: Aloe-emodin, 3-Hydroxymethylchrysazine, 1,8-Dihydroxy-3-(hydroxymethyl)anthracene-9,10-dione, 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone

11677-10 11677-100 11677-25 11677-50
CAYM11677-10EA 65.5 CHF
CAYM11677-10 CAYM11677-100 CAYM11677-25 CAYM11677-50
Aloe emodin 95, crystalline solid
Aloe emodin

Aloe-emodin is a hydroxyanthraquinone found in aloe vera leaves that has potent laxative action by inducing chloride secretion in colonic mucosa and releasing acetylcholine, which stimulates contraction of intestinal smooth muscle.


It also demonstrates anti-tumor activity (ED₅₀s=1 to 13 µM), inducing apoptosis in various cancer cells by increasing the production of reactive oxygen species. Aloe-emodin is reported to have estrogenic activity as a phytoestrogen and has been shown to inhibit breast cancer cell proliferation by downregulating ERα protein levels, and thus, suppressing transcriptional activation of the receptor.

Formel: C₁₅H₁₀O₅
MW: 270.24 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00017373
CAS-Nummer: 481-72-1

Order Now

Spezifikation Testergebnisse

Purity ≥95%
Formulation A Crystalline Solid
λmax 225, 255, 285, 430 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR