Anti-IP6K2 Mouse Monoclonal Antibody [clone: 4F10]

Lieferant: Cayman Chemical
10239-1
CAYM10239-1EA 637 CHF
CAYM10239-1
Anti-IP6K2 Mouse Monoclonal Antibody [clone: 4F10]
Antikörper
Type: Primary
Antigen: IP6K2
Clonality: Monoclonal
Clone: 4F10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR