Anti-dynamin Sheep Polyclonal Antibody

Lieferant: Cayman Chemical
10009786-1
CAYM10009786-1EA 983 CHF
CAYM10009786-1
Anti-dynamin Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: dynamin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR