Piceatannol

Lieferant: Cayman Chemical

Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

10009366-10 10009366-25 10009366-5 10009366-50
CAYM10009366-10EA 180 CHF
CAYM10009366-10 CAYM10009366-25 CAYM10009366-5 CAYM10009366-50
Piceatannol
Piceatannol
Formel: C₁₄H₁₂O₄
MDL Number: MFCD00221715
CAS-Nummer: 10083-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR