Ethyllaurat

Lieferant: Cayman Chemical

Synonyms: Dodecanoic acid ethyl ester, Ethyl dodecanoate, Ethyl Laurate, Dodecylic acid ethyl ester, Lauric acid ethyl ester

10008203-100 10008203-500 10008203-1
CAYM10008203-100EA 30.5 CHF
CAYM10008203-100 CAYM10008203-500 CAYM10008203-1
Ethyllaurat
Ethyllaurat
Formel: C₁₄H₂₈O₂
MW: 228.38 g/mol
Siedepunkt: 269
Melting Pt: –2 °C
Dichte: 0.86 g/cm³ (20 °C)
Flash-Pt: 138
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00015065
CAS-Nummer: 106-33-2
EINECS: 203-386-0
Merck Index: 13,03851

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR