Benzophenon-[Carbonyl-13C] Isotope_(98% 13C)

Lieferant: CAMBRIDGE ISOTOPE LABS

CLM-3253-0.5
CAMBCLM-3253-0.5EA 0 CHF
CAMBCLM-3253-0.5
Benzophenon-[Carbonyl-13C] Isotope_(98% 13C)
Benzophenon-[Carbonyl-13C]
Formel: C₁₃H₁₀O
CAS-Nummer: 32488-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR