Pipette tips, PreCision Line
Lieferant: BIOZYM
760001 760003 760005 760008 760013 760201 760203 760205 760211 760213 760215 760218 761001 761003 761005 761011 761013 761018
BZYM760001EA 0 CHF
BZYM760001 BZYM760003 BZYM760005 BZYM760008 BZYM760013 BZYM760201 BZYM760203 BZYM760205 BZYM760211 BZYM760213 BZYM760215 BZYM760218 BZYM761001 BZYM761003 BZYM761005 BZYM761011 BZYM761013 BZYM761018
Pipette tips, PreCision Line
Pipettenspitzen
Transparent, yellow or blue; sterile or non sterile.
  • Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
  • Autoclavable
  • Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Zertifizierungen: DIN 12650
Order Now
TECHNISCHE DATEN
Environmentally Preferable