Anti-GRB7 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7805
BWRLBS7805EA 488 CHF
BWRLBS7805
Anti-GRB7 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human GRB7.

GRB7 pAb detects endogenous levels of GRB7 protein.

Type: Primary
Antigen: GRB7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR