Anti-CABP3 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS60125
BWRLBS60125EA 375 CHF
BWRLBS60125
Anti-CABP3 Rabbit Polyclonal Antibody
Antikörper
A synthetic peptide corresponding to residues in Human CABP3.

CABP3 pAb detects endogenous levels of CABP3 protein.

Type: Primary
Antigen: CABP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR