Anti-p-Csk Rabbit Polyclonal Antibody [clone: S364]

Lieferant: Bioworld Technology
BS4673
BWRLBS4673EA 406 CHF
BWRLBS4673
Anti-p-Csk Rabbit Polyclonal Antibody [clone: S364]
Antikörper
Synthetic phosphopeptide derived from human Csk around the phosphorylation site of Serine 364.

p-Csk (S364) pAb detects endogenous levels of Csk protein when phosphorylated at Ser364.

Type: Primary
Antigen: p-Csk
Clonality: Polyclonal
Clone: S364
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

TECHNISCHE DATEN

Antigen-Symbol p-Csk
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Klon S364
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Isotyp
Western blot Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR