Anti-p-PKC alpha Rabbit Polyclonal Antibody [clone: T638]

Lieferant: Bioworld Technology
BS4346
BWRLBS4346EA 406 CHF
BWRLBS4346
Anti-p-PKC alpha Rabbit Polyclonal Antibody [clone: T638]
Antikörper
Synthetic phosphopeptide derived from human PKC alpha around the phosphorylation site of Threonine 638.

p-PKC á/â2 (T638) pAb detects endogenous levels of PKC á/â2 protein when phosphorylated at Thr638.

Type: Primary
Antigen: p-PKC alpha
Clonality: Polyclonal
Clone: T638
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR