Anti-p-MEK2 Rabbit Polyclonal Antibody [clone: T394]

Lieferant: Bioworld Technology
BS4122
BWRLBS4122EA 406 CHF
BWRLBS4122
Anti-p-MEK2 Rabbit Polyclonal Antibody [clone: T394]
Antikörper
Synthetic phosphopeptide derived from human MEK2 around the phosphorylation site of Threonine 394.

p-MEK2 (T394) pAb detects endogenous levels of MEK2 protein only when phosphorylated at Thr394

Type: Primary
Antigen: p-MEK2
Clonality: Polyclonal
Clone: T394
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR