Anti-LAT Rabbit Polyclonal Antibody [clone: D156]

Lieferant: Bioworld Technology
BS3757
BWRLBS3757EA 375 CHF
BWRLBS3757
Anti-LAT Rabbit Polyclonal Antibody [clone: D156]
Antikörper
Synthetic peptide, corresponding to amino acids 131-180 of Human LAT.

LAT (D156) pAb detects endogenous levels of LAT protein.

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone: D156
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR