Anti-BMP-8A Rabbit Polyclonal Antibody [clone: Q299]

Lieferant: Bioworld Technology
BS3092
BWRLBS3092EA 375 CHF
BWRLBS3092
Anti-BMP-8A Rabbit Polyclonal Antibody [clone: Q299]
Antikörper
Synthetic peptide, corresponding to amino acids 264-321 of Human BMP-8A.

BMP-8A (Q299) pAb detects endogenous levels of BMP-8A protein. This antibody may cross-reacts with BMP-8B protein.

Type: Primary
Antigen: BMP-8A
Clonality: Polyclonal
Clone: Q299
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR