Rotary evaporator, large scale, Rotavapor® R-250 EX
Lieferant: BUCHI
8610C511 8610C512 8610C521 8610C522 8610C541 8610C542 8610C551 8610C552 8610D511 8610D512 8610D521 8610D522 8610D541 8610D542 8610D551 8610D552 8610G511 8610G512 8610G521 8610G522 8610G541 8610G542 8610G551 8610G552 8611C511 8611C512 8611C521 8611C522 8611C541 8611C542 8611C551 8611C552 8611D511 8611D512 8611D521 8611D522 8611D541 8611D542 8611D551 8611D552 8611G511 8611G512 8611G521 8611G522 8611G541 8611G542 8611G551 8611G552 8620C511 8620C512 8620C521 8620C522 8620C541 8620C542 8620C551 8620C552 8620D511 8620D512 8620D521 8620D522 8620D541 8620D542 8620D551 8620D552 8620G511 8620G512 8620G521 8620G522 8620G541 8620G542 8620G551 8620G552 8621C511 8621C512 8621C521 8621C522 8621C541 8621C542 8621C551 8621C552 8621D511 8621D512 8621D521 8621D522 8621D541 8621D542 8621D551 8621D552 8621G511 8621G512 8621G521 8621G522 8621G541 8621G542 8621G551 8621G552
BUCH8610C511EA 0 CHF
BUCH8610C511 BUCH8610C512 BUCH8610C521 BUCH8610C522 BUCH8610C541 BUCH8610C542 BUCH8610C551 BUCH8610C552 BUCH8610D511 BUCH8610D512 BUCH8610D521 BUCH8610D522 BUCH8610D541 BUCH8610D542 BUCH8610D551 BUCH8610D552 BUCH8610G511 BUCH8610G512 BUCH8610G521 BUCH8610G522 BUCH8610G541 BUCH8610G542 BUCH8610G551 BUCH8610G552 BUCH8611C511 BUCH8611C512 BUCH8611C521 BUCH8611C522 BUCH8611C541 BUCH8611C542 BUCH8611C551 BUCH8611C552 BUCH8611D511 BUCH8611D512 BUCH8611D521 BUCH8611D522 BUCH8611D541 BUCH8611D542 BUCH8611D551 BUCH8611D552 BUCH8611G511 BUCH8611G512 BUCH8611G521 BUCH8611G522 BUCH8611G541 BUCH8611G542 BUCH8611G551 BUCH8611G552 BUCH8620C511 BUCH8620C512 BUCH8620C521 BUCH8620C522 BUCH8620C541 BUCH8620C542 BUCH8620C551 BUCH8620C552 BUCH8620D511 BUCH8620D512 BUCH8620D521 BUCH8620D522 BUCH8620D541 BUCH8620D542 BUCH8620D551 BUCH8620D552 BUCH8620G511 BUCH8620G512 BUCH8620G521 BUCH8620G522 BUCH8620G541 BUCH8620G542 BUCH8620G551 BUCH8620G552 BUCH8621C511 BUCH8621C512 BUCH8621C521 BUCH8621C522 BUCH8621C541 BUCH8621C542 BUCH8621C551 BUCH8621C552 BUCH8621D511 BUCH8621D512 BUCH8621D521 BUCH8621D522 BUCH8621D541 BUCH8621D542 BUCH8621D551 BUCH8621D552 BUCH8621G511 BUCH8621G512 BUCH8621G521 BUCH8621G522 BUCH8621G541 BUCH8621G542 BUCH8621G551 BUCH8621G552
Rotary evaporator, large scale, Rotavapor® R-250 EX
Verdampfer Rotationsverdampfer
The Rotavapor R-250 EX is the EX rated version of the Rotavapor R-250. It fulfills the high requirements of the ATEX regulations and fits perfect to your Ex-proof production laboratory. It is designed for easy and safe operation combined with high performance.
  • Intuitive operation and handling guarantee safe and efficient industrial distillation
  • Immediate shut-down due to safety cut-off
  • Safe handling and spring-loaded valves protect the glass
  • Easy flask handling due to the unique flask coupling
  • Long life time due to exclusive use of corrosion-free stainless
More Product Information