Anti-AMHR2 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PB9984
BSBTPB9984EA 696 CHF
BSBTPB9984
Anti-AMHR2 Rabbit Polyclonal Antibody
Antikörper
AMHR2 is the receptor for the anti-Mullerian hormone (AMH) which, in addition to testosterone, results in male sex differentiation. AMH and testosterone are produced in the testes by different cells and have different effects. Testosterone promotes the development of male genitalia while the binding of AMH to the encoded receptor prevents the development of the mullerian ducts into uterus and Fallopian tubes. Mutations in this gene are associated with persistent Mullerian duct syndrome type II. Alternatively spliced transcript variants encoding different isoforms have been identified.