Anti-BAFF Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PB9567
BSBTPB9567EA 696 CHF
BSBTPB9567
Anti-BAFF Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Tumor necrosis factor ligand superfamily member 13B (TNFSF13B) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: BAFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR