Anti-WNT1 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
PB9460
BSBTPB9460EA 696 CHF
BSBTPB9460
Anti-WNT1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Proto-oncogene Wnt-1(WNT1) detection. Tested with WB in Human.
Wingless-type MMIV integration site family, member 1 is a protein that in humans is encoded by the WNT1 gene. This gene is a member of the WNT gene family. The gene was assigned to human chromosome 12q13.12. The WNT gene family consists of structurally related genes that encode secreted signaling proteins. The Wnt1 protein functions in the induction of the mesencephalon and cerebellum. However, the gene mutations might not have a significant role in Joubert syndrome.
Add 0.2ml of distilledMore Product Information