Anti-GRIA3 Polyclonal Antibody

Lieferant: Boster Bio
PB9206
BSBTPB9206EA 696 CHF
BSBTPB9206
Anti-GRIA3 Polyclonal Antibody
Antikörper
Polyclonal antibody for GLUR 3/GRIA3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. GLUR 3/GRIA3 information: Molecular Weight: 101157 MW; Subcellular Localization: Cell membrane; Multi-pass membrane protein. Cell junction, synapse, postsynaptic cell membrane; Multi-pass membrane protein. Interaction with CNIH2 and CNIH3 promotes cell surface expression.

Type: Primary
Antigen: GRIA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR