Anti-Aquaporin 0 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA2110
BSBTPA2110EA 696 CHF
BSBTPA2110
Anti-Aquaporin 0 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Lens fiber major intrinsic protein(MIP) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Aquaporin 0
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR