Anti-ALDH3A1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA2074
BSBTPA2074EA 696 CHF
BSBTPA2074
Anti-ALDH3A1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Aldehyde dehydrogenase, dimeric NADP-preferring(ALDH3A1) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: ALDH3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR