Anti-E2F6 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1811
BSBTPA1811EA 696 CHF
BSBTPA1811
Anti-E2F6 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Transcription factor E2F6 (E2F6) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: E2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR