Anti-ABCB6 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1723
BSBTPA1723EA 696 CHF
BSBTPA1723
Anti-ABCB6 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for ATP-binding cassette sub-family B member 6, mitochondrial (ABCB6) detection. Tested with WB, IHC-P, ICC in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: ABCB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR