Anti-CD68 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1518
BSBTPA1518EA 696 CHF
BSBTPA1518
Anti-CD68 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Macrosialin(CD68) detection. Tested with WB, IHC-P, IHC-F in Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CD68
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR