Anti-KCNN4 Rabbit Antibody

Lieferant: Boster Bio
PA1047-1
BSBTPA1047-1EA 696 CHF
BSBTPA1047-1
Anti-KCNN4 Rabbit Antibody
Antikörper
Rabbit IgG polyclonal antibody for Intermediate conductance calcium-activated potassium channel protein 4(KCNN4) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: KCNN4
Clonality:
Clone:
Conjugation:
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR