Anti-Synaptophysin Mouse Monoclonal Antibody [clone: Syn-58]

Lieferant: Boster Bio
MA1091
BSBTMA1091EA 696 CHF
BSBTMA1091
Anti-Synaptophysin Mouse Monoclonal Antibody [clone: Syn-58]
Antikörper
Mouse IgG monoclonal antibody for Synaptophysin, synaptophysin (SYP) detection. Tested with WB, IHC-F in Human; Rat. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Synaptophysin
Clonality: Monoclonal
Clone: Syn-58
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR