Anti-p34cdc2 Mouse Monoclonal Antibody [clone: IMD-34]

Lieferant: Boster Bio
MA1077
BSBTMA1077EA 696 CHF
BSBTMA1077
Anti-p34cdc2 Mouse Monoclonal Antibody [clone: IMD-34]
Antikörper
Type: Primary
Antigen: p34cdc2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR