Mouse BMP-4 PicoKine; ELISA Kit

Lieferant: Boster Bio

EK0316
BSBTEK0316EA 881 CHF
BSBTEK0316
Mouse BMP-4 PicoKine; ELISA Kit
Tests ELISAs

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse BMP-4.


  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts


Detect Mouse Bmp4 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: bone tissue, cell culture supernates and serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Bmp4 tissue specificity:


96wells/kit, with removable strips.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR