Anti-IgG Rabbit Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Boster Bio
BA1110-0.5
BSBTBA1110-0.5EA 0 CHF
BSBTBA1110-0.5
Anti-IgG Rabbit Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR