Anti-Steptavidin Antibody (Peroxidase)

Lieferant: Boster Bio
BA1088-1
BSBTBA1088-1EA 0 CHF
BSBTBA1088-1
Anti-Steptavidin Antibody (Peroxidase)
Antikörper
Type: Primary
Antigen: Steptavidin
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR