Anti-AP2M1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A06179
BSBTA06179EA 696 CHF
BSBTA06179
Anti-AP2M1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for AP-2 complex subunit mu(AP2M1) detection. Tested with WB in Human, Mouse, Rat.

Type: Primary
Antigen: AP2M1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR