Anti-P2RY5 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A05725-1
BSBTA05725-1EA 696 CHF
BSBTA05725-1
Anti-P2RY5 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for P2RY5 detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: P2RY5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR